Learn More
Static Solutions OHM-Shield AF-6000 Zero VOC ESD Floor Finish
Combines static protection, durability and gloss with environmentally friendly chemicals.
$251.00 - $419.00
Specifications
For Use With (Application) | Medical, electronics and cleanroom environments, this product protects the floor with a long lasting and durable gloss |
---|---|
Product Type | OHM-Shield™ AF-6000 Zero VOC ESD Floor Finish |
Catalog Number | Mfr. No. | Volume (English) | Volume (Metric) | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Volume (English) | Volume (Metric) | Price | Quantity & Availability | ||||
19-130-6032
|
Static Solutions
AF6000 |
1 gal. | 3.8 L |
Case of 4 for $251.00
|
|
19-130-6033
|
Static Solutions
AF6005 |
5 gal. | 18.9 L |
Each for $419.00
|
|
Description
Protects the floor with a long lasting and durable gloss without harming the environment. No buffing required. Produces zero VOCs (Volatile Organic Compounds); eliminates the possibility of outgassing. Safe for your workers as well as your piece parts. Can be used in closed environments.
- High gloss
- Zinc-free
- Biodegradable
- Low odor
- Static dissipative
- No dye or fragrance
- High percentage of solids (18-20%)
Order Info
Resistance: 107 to 109 ohms/sq. Static Dissipation: 0.02 seconds. Color: Translucent white; dries clear. Coverage: 1500-2500 sq. ft. per gallon. Note: Allow 1-2 hours of drying time between coats (2-3 recommended). Coated floor should be earth grounded for rapid dissipation. Store tightly closed in a cool dry area, and do not mix other floor finishes or left over products from previous applications.
Certifications
UL listed for slip resistance.
Compliance
EOS/ESD S202-20 compliant. RoHS compliant. Exceeds EPA guidelines.
Specifications
Medical, electronics and cleanroom environments, this product protects the floor with a long lasting and durable gloss | |
OHM-Shield™ AF-6000 Zero VOC ESD Floor Finish |
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.