Learn More
Static Solutions™ OHM-Shield™ TA-4900 Cleanroom Topical Antistat
Eliminates and prevents static electricity on cleanroom surfaces
Manufacturer:  Static Solutions™ TA4900
Catalog No. 19-130-6034
Add to cart
This item is not returnable. View return policy
Description
Description
Cleans and rejuvenates static dissipative mats, and makes non-dissipative surfaces dissipative.
  • Makes all surfaces static dissipative to less than 1010 ohms
  • Quick-drying and nonstreaking
  • Water-based
  • Filtered to less than 0.2 microns
  • No chlorine, bromine, nitrate, or sulfate
  • Low ion and low hydrocarbon
  • Noncorrosive and nonflammable
Order Info
Static Decay Rate: Less than 0.01 seconds. Translucent. Available in double-bag packaging for cleanroom entry. UBA# 39890009.
Specifications
Specifications
OHM-Shield™ TA-4900 cleanroom topical antistat
32 oz., 0.94L
OHM-Shield™ TA-4900
32 oz.
Designed to control, prevent and eliminate static electricity in surfaces in a cleanroom environment
0.94L