Learn More
Static Solutions™ OHM-Stat™ CT-8900 Combination Tester/Data Logger
SDP
No more log books—tests foot grounder and wrist strap in three seconds
Supplier:  Static Solutions™ CT8950
Encompass
 View more versions of this product
Catalog No. 19-130-6020
Add to Cart
Includes: NIST certificate.
Description
Description
Measures the electrical resistance of each foot and wrist strap simultaneously and individually with one push of a button. Offers adjustable limits, exact resistivity readings, unique report generation groups, versatile networking options, and an expandable employee log of 30,000. Made in USA.
  • Increases worker productivity, reduces defects, and saves administrative time
  • Accepts mag stripe, barcode, Motorola™, HID™, and Casi-Rusco™ proximity ID cards as well as keyboard entry
  • Included ESD Protection Test Log software generates e-mail reports to multiple recipients
  • Logs are easily imported into spreadsheet/database programs
  • Prevents cheating—exception report will inform if worker did not test once or twice per day
  • Able to prevent unauthorized entry without proper ID/ESD certification; can be upgraded to open electronically locked doors with pass of test
  • Measures from 100 kilohms to 100 megohms, with an option to 1000 megohms
  • Tests performed at 19 volts—not competitive 9 volts
Order Info
Compatible with Microsoft™ Windows™ 95, 98, ME, NT, and 2000 operating systems. Can use Oracle™, Access™, or Excel™ databases. Powered by 9 volt battery (IEC6F22) or 12 volt DC adapter.
Compliance
Conforms to ESD-S 20-20, European EN, DIN and military standards. NIST and RoHs compliant. CE listed.
Specifications
Specifications
Combo Tester with Footplate, Stand, and Bar Code or Mag Stripe Reader
Product Suggestions
Product Suggestions