Learn More
Fisherbrand™ Stationary plug - Medical Nylon - QC
SDP
Superior single barb technology that provides a leak-proof seal. Ideal with flexible tubing. Nut and body are locked as a single unit. Precision engineered for smooth positive sealing. Color nut for easy recognition. This standard luer is ISO 80369-7 compliant. 
 View more versions of this product
Catalog No. 01-000-497
Add to cart
Description
Description
  • Single & Multi Use Applications
  • PVC-Free
  • ISO 80369-7 Compliant
  • Superior Single Barb Design
  • USP Class VI Compliant
  • USP 661, USP81, USP 88
  • RoHS Compliant
  • Gamma Stable
  • Animal Derivative Free
  • Lot Traceable
Warranty and Services
Replacement if not satisfied. 12 Month return Policy.
Specifications
Specifications
Compliance Certificates Available
Natural Nylon
Tubing connections; Medical device input/output ports; Drug and fluid transfer lines
Out of direct sunlight, >20°F <100°F
Class 7 Cleanroom Manufactured
Stationary Luer Lock Plug
100 Pack
Violet Purple