Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Our pricing system is unavailable. You are viewing list price.
Please call 1-800-766-7000 to place your order or try our site again later.

Fisherbrand™ Stationary plug - Polypropylene - QC

Superior single barb technology that provides a leak-proof seal. Ideal with flexible tubing. Nut and body are locked as a single unit. Precision engineered for smooth positive sealing. Color nut for easy recognition. This standard luer is ISO 80369-7 compliant. 

$63.30 - $66.50

Specifications

Material Non-Animal Derived Polypropylene
Type Stationary Luer Lock Plug
For Use With (Application) Tubing connections; Medical device input/output ports; Drug and fluid transfer lines
Description Class 7 Cleanroom Manufactured
Certifications/Compliance Compliance Certificates Available
View More Specs

Products 7
Catalog Number Color Price Quantity & Availability  
Catalog Number Color Price Quantity & Availability  
01-000-499 View Documents Berry Blue Pack of 100 for $66.50
N/A
01-000-500 View Documents Lime Green Pack of 100 for $66.50
N/A
01-000-501 View Documents Natural Pack of 100 for $63.30
N/A
01-000-503 View Documents Rose Red Pack of 100 for $66.50
N/A
01-000-502 View Documents Orange Pack of 100 for $66.50
N/A
01-000-504 View Documents Violet Purple Pack of 100 for $66.50
N/A
01-000-505 View Documents Yellow Pack of 100 for $66.50
N/A
Description

Description

  • Single & Multi Use Applications
  • PVC-Free
  • ISO 80369-7 Compliant
  • Superior Single Barb Design
  • USP Class VI Compliant
  • USP 661, USP81, USP 88
  • RoHS Compliant
  • Gamma Stable
  • Animal Derivative Free
  • Lot Traceable

Warranty and Services

Replacement if not satisfied. 12 Month return Policy.

Specifications

Specifications

Non-Animal Derived Polypropylene
Tubing connections; Medical device input/output ports; Drug and fluid transfer lines
Compliance Certificates Available
Stationary Luer Lock Plug
Class 7 Cleanroom Manufactured
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit