Learn More
HemoCue America StatSpin Blood Separation Prepared Microtubes
Catalog Number | Mfr. No. | Description | Price | Qty | Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Description | Price | Qty | Availability | ||||
22-269-523
|
HemoCue America
TP1U |
N/A | 1.3mL microtube; Serum; With clot activator |
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. |
N/A |
Description
- Serum (red top), plasma (green top), or gel tubes (clear top)
- Separations in only 30 to 90 seconds
- Fit RT 12 tube rotor, Cat. No. 22-269-258
- For use with StatSpin VT Veterinary Multipurpose Centrifuges 22-269-520, -521
- May also be used with the StatSpin MP (22-314-934), RP (22-314-935) and CritSpin (22-269-291, -292, -294, 22-305-276) centrifuges
Specifications
Plastic |