Learn More
Beckman Coulter™ StatSpin™ Digital Hematocrit Reader
Increase the productivity of any StatSpin centrifuge with hematocrit capability. With the StatSpin Digital Hematocrit Reader, an easy-to-use pointer and bright illumination made reading tubes simple.
Manufacturer: Beckman Coulter™ CSD2
Description
- Can be used with CritSpin or MP models
- Digital reader with large HCT display simplifies tube reading and minimizes error
- Readings are available as percent (to 0.5%) or decimal equivalent (to 0.005)
- Readings are unaffected by tube diameter, size or material
- Accepts RH12 rotor directly with no need to transfer tubes
- Large, easy-to-use pointer
- Brightly illuminated face
- 75mm tube adapter allows unit to work with other systems
Specifications
Power supply | |
Digital Hematocrit Reader |