Learn More
  Our pricing system is currently unavailable. You are viewing list price.
You can save item(s) to your list now to purchase when our pricing system is available.
StatSpin™ Glass Microhematocrit Tubes
Manufacturer:  HemoCue America HT9U10
 View more versions of this product
Catalog No. 22-269246
Add to cart
Specifications Description & Specifications
Specifications
Anti-coagulated samples
1.57 in.
0.0003 oz.
Glass
RH12 HCT rotor
4cm
9μL
Microhematocrit Capillary Tube
Safety and Handling
ShelfLife : 2 Year(s)