Learn More
HemoCue America StatSpin™ LipoClear™ Tubes Available on GSA/VA Contract for Federal Government customers only.
Clear lipemic serum or plasma with any standard centrifuge: no ultracentrifuge or organic solvents needed
Manufacturer:  HemoCue America LCP1
 View more versions of this product
Catalog No. 22-274-916
Add to cart
This item is not returnable. View return policy
Description
Description
Clear lipemic serum or plasma with any standard centrifuge: no ultracentrifuge or organic solvents needed
  • LipoClear tubes are pre-filled and ready-to-use with a nontoxic, non-carcinogenic reagent
  • Clearing time varies with centrifuge model (only 95 seconds with StatSpin-MP)
  • LipoClearPlus in 30mL size available for higher volume laboratories
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Specifications
Clears lipemic samples in 95sec. with a StatSpin™ LipoClear™ centrifuge tube
Mix 1 part LipoClear™ Plus to 5 parts patient sample
2° to 8°C
Clearing reagent
StatSpin™ LipoClear™ high speed centrifuge and other standard centrifuge
30mL
95 sec.
Safety and Handling
Safety and Handling
ShelfLife : 1 year