Learn More
HemoCue America StatSpin™ LipoClear™ Tubes
Clear lipemic serum or plasma with any standard centrifuge: no ultracentrifuge or organic solvents needed
Manufacturer:  HemoCue America LC15
 View more versions of this product
Catalog No. 22-287646
Add to cart
This item is not returnable. View return policy
Description
Description
Clear lipemic serum or plasma with any standard centrifuge: no ultracentrifuge or organic solvents needed
  • LipoClear tubes are pre-filled and ready-to-use with a nontoxic, non-carcinogenic reagent
  • Clearing time varies with centrifuge model (only 95 seconds with StatSpin-MP)
  • LipoClearPlus in 30mL size available for higher volume laboratories
Specifications
Specifications
Blood collection
1.5mL (40/Pk.)
2° to 8°C
Tube rotor, can also be used in StatSpin™ LipoClear™ high speed centrifuge and other standard centrifuge
0.3mL
Microcentrifuge capillary tube
Safety and Handling
Safety and Handling
ShelfLife : 1 year