Learn More
HemoCue America StatSpin™ Sealant Pad for Capillary Tubes
Manufacturer:  HemoCue America HS2410
Catalog No. 22-269247
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Description
Description
  • Clay-like material used for sealing microhematocrit tubes
  • Includes holder tray with 24 numbered slots for tube storage
Specifications
Specifications
Used for sealing microhematocrit capillary tube
Holder tray with 24 numbered slots
10 Pack
Sealant Pad
CritSpin™ microhematocrit capillary tube
24.0
Clay