StatSpin™ StatSampler™ Blood Collection System
Combines a capillary tube for collection and a microtube for sampling
Manufacturer: Beckman Coulter Primary Care Diagnostics SS2E
 View all options for this product
Catalog No. 22-282471
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. 
    Capacity (Metric) 200μL
    Additive EDTA
    Color Lavender
    Product Type Tube
    • Capillary tube draws blood quickly via capillary action (no scooping necessary)
    • Immediate anticoagulation reduces microclots—makes EDTA variation perfect for hematology analyzers
    • Heparin-treated and untreated collection systems for plasma/serum separation and chemistry analysis
    • Both systems have a gel barrier
    • Spin in a StatSpin centrifuge or any benchtop or floor model centrifuge capable of spinning microtubes
  • Description & Specifications
Related Products
Product Content Feedback