Learn More
LifeSign Status BTA Urine Test Kit
Status BTA is a rapid test for the qualitative detection of bladder tumor associated antigen in human urine.
Specifications
Detectable Analytes Bladder tumor associated antigen
Test Time 5 minutes
View More Specs
Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
23-264-797
View Documents
LifeSign
39010
10 tests
Pack of 10 for $199.88
N/A
Due to product restrictions, please to purchase or view availability for this product.
23-264-798
View Documents
LifeSign
39025
25 tests
Pack of 25 for $482.78
N/A
Due to product restrictions, please to purchase or view availability for this product.
Description
Description
The test is indicated for use as an aid in the management of bladder cancer patients in conjunction with cystoscopy. The simple urine sample collection also allows for improved patient comfort. Because tumor markers can be used to assess the response of a tumor treatment and for prognosis, rapid screening tests like the Status BTA product are very helpful in the treatment management of cancer patients.
  • CLIA Waived
  • Results available in 5 minutes
  • Simple procedure requires only adding 3 drops of urine to test device; no sample preparation needed
  • Accurate test results with excellent sensitivity and specificity
  • Uses monoclonal antibodies to detect the presence of bladder tumor associated antigen in urine
  • Detects early state recurrance cancer that cytology may miss
  • Adjunct test to cystoscopy
The test is indicated for use as an aid in the management of bladder cancer patients in conjunction with cystoscopy.  The simple urine sample collection also allows for improved patient comfort.  Because tumor markers can be used to assess the response of a tumor treatment and for prognosis, rapid screening tests like the Status BTA product are very helpful in the treatment management of cancer patients.
  • CLIA Waived
  • Results available in 5 minutes
  • Simple procedure requires only adding 3 drops of urine to test device; no sample preparation needed
  • Accurate test results with excellent sensitivity and specificity
  • Uses monoclonal antibodies to detect the presence of bladder tumor associated antigen in urine
  • Detects early state recurrance cancer that cytology may miss
  • Adjunct test to cystoscopy
Specifications
Specifications
Bladder tumor associated antigen
5 minutes