Learn More
LifeSign Status DS 8 Panel
One-step immunoassay for the qualitative detection of drugs of abuse and their metabolites in urine. Accurate results with a 97 to 99% correlation to GC/MS.
Manufacturer:  LifeSign 28025
Catalog No. 23-264-779
Add to cart
Description
Description
  • Marijuana (THC)/Opiates (OPI)/Cocaine (COC)/Amphetamine (AMP)/Benzodiazepines (BZO)/Barbiturates (BAR)/Tricyclic Antidepressants (TCA)/Phencyclidine (PCP)
  • Simple one-step procedure
  • Fast turnaround time with results in 5 to 10 minutes
  • Flexible combination of drugs from a single drug up to 10 drug tests
Specifications
Specifications
25 Tests