Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ STBD1 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA552993
Description
Immunogen sequence: APVLNLNQGM DNGRSTLVEA RGQQVHGKME RVAVMPAGSQ QVSVRFQVHY VTSTDVQFIA VTGDHECLGR WNTYIPLHYN KDGFWSHSIF LPADTVVEWK FVLVENGGVT RWEECSNRFL ETGHEDKVVH AWW Highest antigen sequence identity to the following orthologs: Mouse - 71%, Rat - 72%.
Acts as a cargo receptor for glycogen. Delivers its cargo to an autophagic pathway called glycophagy, resulting in the transport of glycogen to lysosomes. [UniProt].
Specifications
STBD1 | |
Polyclonal | |
Unconjugated | |
STBD1 | |
D530019K15Rik; D5Ertd593e; genethonin 1; genethonin-1; GENEX3414; GENX-3414; Glycophagy cargo receptor stbd1; RGD1311800; starch binding domain 1; starch-binding domain-containing protein 1; Stbd1 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
8987 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin) | |
0.1 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
O95210 | |
STBD1 | |
Recombinant protein corresponding to Human STBD1. Recombinant protein control fragment (Product #RP-90146). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction