Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

STC1 Mouse anti-Canine, Human, Clone: OTI6D12, Invitrogen™

Mouse Monoclonal Antibody

Manufacturer:  Invitrogen MA527246

Catalog No. PIMA527246


Add to cart

Description

Description

This gene encodes a secreted, homodimeric glycoprotein that is expressed in a wide variety of tissues and may have autocrine or paracrine functions. The gene contains a 5 ft. UTR rich in CAG trinucleotide repeats. The encoded protein contains 11 conserved cysteine residues and is phosphorylated by protein kinase C exclusively on its serine residues. The protein may play a role in the regulation of renal and intestinal calcium and phosphate transport, cell metabolism, or cellular calcium/phosphate homeostasis. Overexpression of human stanniocalcin 1 in mice produces high serum phosphate levels, dwarfism, and increased metabolic rate. This gene has altered expression in hepatocellular, ovarian, and breast cancers.
Specifications

Specifications

STC1
Monoclonal
1 mg/mL
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3
P52823
Stanniocalcin-1 [Precursor]; STC-1; STC1; STC
Mouse
IgG1
100 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
486112, 6781
Western Blot
OTI6D12
Unconjugated
STC1
Liquid
STC1
Human recombit protein fragment corresponding to amino acids 34-247 of STC1 produced in E.coli
Affinity Chromatography
RUO
Antibody
Monoclonal
Canine, Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit