Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Pelican™ StealthLite™ Flashlight and Rechargeable System

IPX7 rating

Supplier:  Pelican™ 2400CLG

 View more versions of this product

Catalog No. 19-822-650

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Includes: Battery-Powered System: Four “AA” alkaline batteries.
Rechargeable System: StealthLite flashlight, desk/dash mount base, 110V charger, and high-capacity NiMH battery pack.

Description

Description

  • 3.75 hours burn time
  • Lexan EXL resin
  • Black lanyard

Battery-Powered System

  • Xenon Laser Spot lamp module with white collimated beam One-hand safety switch and double-shock-mounted battery compartment
Rechargeable System
    High-performance Xenon Laser Spot lamp module has 50% higher candlepower than module for battery-powered StealthLite

Order Info

Length: 6.5 in. (16.5cm). Weight (with batteries): 7.1 oz. (201g).

Certifications

CSA and FM approved for certain Class I, Division 2 hazardous locations; ETL marked.

Specifications

Specifications

Flashlight
6 V
0.2 kg
Alkaline
Lime Green
4 AA alkaline batteries
16.48 cm
EXL Resin
Xenon
7.1 oz.
3.45 hr.
CSA, FM, Class I, Division 2, ETL, ANSI/NEMA FL 1
The tough resin body is corrosion proof and resistant to extreme temperatures, xenon laser spot lamp module with white collimated beam one-hand safety switch and double shock mounted battery compartment
6.49 in.
45 lm
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit