Learn More
Electron Microscopy SciencesSupplier Diversity Partner Steam Autoclave Printed Tape (B), 3/4" wide, 3" core 60 YD DFS Item
Manufacturer:  Electron Microscopy SciencesSupplier Diversity Partner 7701707
Indicate the status of all laboratory items to be sterilized with these tape labels. The word "STERILIZED" appears after processing at 250 degree F under normal sterilization conditions.
Width: 3/4"
Core: 3" core
Catalog No. 50-253-96