Learn More
Labconco™ SteamScrubber™ and FlaskScrubber™ Culture Tube Insert
SCI_ED
For use with SteamScrubber™ and FlaskScrubber™ laboratory glassware washers. Labconco™ SteamScrubber™ and FlaskScrubber™ Culture Tube Inserts hold tubes for easy transport and safe washing and drying.
Specifications
Type Culture Tube Insert
Width (Metric) 45.7cm
For Use With (Equipment) SteamScrubber and FlaskScrubber Glassware Washer
Material Type 304 stainless steel
View More Specs
Products 3
Catalog Number Mfr. No. Capacity Description Size Height (Metric) Price Quantity & Availability  
Catalog Number Mfr. No. Capacity Description Size Height (Metric) Price Quantity & Availability  
S28368
SCI_ED
View Documents Promotion Details
Labconco™
4588600/EMD
240 tubes Holds: 10 - 12mm tubes 16.8 x 16.8mm 11.4cm This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000. N/A
S28376
SCI_ED
View Documents Promotion Details
Labconco™
4402001/EMD
144 tubes Holds: 15 - 18mm tubes 23.8 x 23.8mm 16.2cm This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000. N/A
S28377
SCI_ED
View Documents Promotion Details
Labconco™
4402101/EMD
105 tubes Holds: 20 - 24mm tubes 26.5 x 26.5mm 25.1cm This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000. N/A
Description
Description
  • Safely and easily wash and dry culture tubes
  • Hold tubes for easy transport and safe washing and drying
  • Type 304 stainless steel
  • May be placed in upper or lower Standard Open or Spindle Rack in Labconco FlaskScrubber or SteamScrubber
Warranty and Services
One year
Specifications
Specifications
Culture Tube Insert
45.7cm
SteamScrubber and FlaskScrubber Glassware Washer
Type 304 stainless steel