Learn More
STEAP2 Rabbit anti-Human, Mouse, Polyclonal, Proteintech Non-distribution product as customer accommodation.
Rabbit Polyclonal Antibody
Manufacturer:  Proteintech Group IncSupplier Diversity Partner 202011AP
Catalog No. 50-173-2766
Add to cart
Description
Description
The antibody is specific to STEAP2.
The six-transmembrane epithelial antigen of prostate 2 (STEAP2) is a member of a family of metalloreductases identified as cell-surface antigens in prostate tissue. Similar to two other members of the STEAP family (STEAP 3 and STEAP4), STEAP2 promotes both iron and copper reduction. STEAP2 expression in transfected cells also correlated with iron or copper uptake, suggesting that the STEAP family of proteins may function to stimulate iron and copper uptake. STEAP2 is widely expressed in many tissues in the plasma membrane, but is most highly expressed in prostate. At least three isoforms of STEAP2 are known to exist.
Specifications
Specifications
STEAP2
0.28 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Q8BWB6, Q8NFT2
IPCA1, Metalloreductase STEAP2, PCANAP1, PUMPCn, STAMP1, STEAP2, STMP
Rabbit
IgG
150 μL
-20°C
Primary
261729, 74051
Immunohistochemistry (Paraffin), Western Blot
Unconjugated
STEAP2
Liquid
Steap2
Peptide
Antigen Affinity Chromatography
RUO
Antibody
Polyclonal
Human, Mouse