Learn More
STEAP3 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech Non-distribution product as customer accommodation.
Rabbit Polyclonal Antibody
Manufacturer:  Proteintech Group IncSupplier Diversity Partner 171861AP
Catalog No. 50-173-1430
Add to cart
Description
Description
The six-transmembrane epithelial antigen of prostate 3 (STEAP3) is a member of a family of metalloreductases identified as cell-surface antigens in prostate tissue. Similar to two other members of the STEAP family (STEAP 2 and STEAP4), STEAP3 promotes both iron and copper reduction. STEAP3 is highly expressed in hematopoietic tissues and colocalizes with the transferrin endosome. Overexpression of STEAP3 stimulates iron reduction; mice lacking STEAP3 are deficient in erythroid ferrireductase activity, suggesting that STEAP3 is an endosomal ferrireductase required for transferrin-dependent iron uptake in erythroid cells.
Specifications
Specifications
STEAP3
0.28 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Q5RKL5, Q658P3, Q8CI59
dudlin 2, Dudulin 2, hpHyde, hTSAP6, Metalloreductase STEAP3, pHyde, STEAP family member 3, STEAP3, STMP3, TSAP6
Rabbit
IgG
150 μL
-20°C
Primary
170824, 55240, 68428
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
Unconjugated
STEAP3
Liquid
Steap3
STEAP3 Fusion Protein Ag10970
Antigen Affinity Chromatography
RUO
Antibody
Polyclonal
Human, Mouse, Rat