Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Stearic acid, 97%, ACROS Organics™

Click to view available options
Quantity:
1kg
2.5kg
10kg
Packaging:
Fibre drum
Plastic Bottle
Plastic drum
57-11-4
C18H36O2
284.48
MFCD00002752
QIQXTHQIDYTFRH-UHFFFAOYSA-N
stearic acid, stearophanic acid, n-octadecanoic acid, cetylacetic acid, pearl stearic, stearex beads, octadecansaeure, stearinsaeure, vanicol, 1-heptadecanecarboxylic acid
5281
CHEBI:28842
octadecanoic acid
CCCCCCCCCCCCCCCCCC(O)=O
57-11-4
C18H36O2
284.48
MFCD00002752
QIQXTHQIDYTFRH-UHFFFAOYSA-N
stearic acid, stearophanic acid, n-octadecanoic acid, cetylacetic acid, pearl stearic, stearex beads, octadecansaeure, stearinsaeure, vanicol, 1-heptadecanecarboxylic acid
5281
CHEBI:28842
octadecanoic acid
CCCCCCCCCCCCCCCCCC(O)=O
Sublimation Point >90 °C
Infrared Spectrum Authentic
Flash Point 196°C
Density 0.8400g/mL
Quantity 10kg
Packaging Fibre drum
Specific Gravity 0.84
Color White
Melting Point 67.0°C to 69.0°C
Boiling Point 361.0°C
Assay Percent Range 96% min. (GC)
Linear Formula CH3(CH2)16CO2H
Beilstein 02,IV,1206
Merck Index 15,893
Solubility Information Solubility in water: slightly soluble. Other solubilities: 1g/21mL alcohol- 1g/6mL ccl4,1g/5mL benzene- 1g/3.4mL cs2,1g/2mL chloroform - 1g/26mL acetone,soluble in amyl acetate and toluene
Formula Weight 284.47
Physical Form Granular Powder, Granules, or Flakes
Percent Purity 97%
Chemical Name or Material Stearic acid
Show More Show Less

EINECSNumber : 200-313-4