Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Stearic Acid, Type 50, Powder, NF, 40-60%, Spectrum™ Chemical

Click to view available options
Quantity:
500 g
2.5 kg
12 kg
45 kg
Packaging:
Poly Pail
Description
Spectrum™ Chemical Stearic Acid, Type 50, Powder, NF is used as an emulsifying agent, solubilizing agent, tablet and capsule lubricant. The NF grade indicates it is graded suitable for personal care, cosmetic and pharmaceutical applications. All Spectrum Chemical NF grade products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.
Specifications
Specifications
CAS Max % | 1 |
Boiling Point | 360°C |
Ignition Residue | 0.001 |
Packaging | Poly Pail |
Quantity | 45 kg |
Percent Purity | 40 to 60% |
Grade | NF |
Physical Form | Crystalline Powder or Lumps |
Chemical Name or Material | Stearic Acid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction