Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Stearic Anhydride, Spectrum™

Click to view available options
Quantity:
5 g
25 g
100 g
Packaging:
Amber Glass Bottle
638-08-4
C36H70O3
550.95
WVJVHUWVQNLPCR-UHFFFAOYSA-N
octadecanoyl octadecanoate
CCCCCCCCCCCCCCCCCC(=O)OC(=O)CCCCCCCCCCCCCCCCC
638-08-4
C36H70O3
550.95
WVJVHUWVQNLPCR-UHFFFAOYSA-N
octadecanoyl octadecanoate
CCCCCCCCCCCCCCCCCC(=O)OC(=O)CCCCCCCCCCCCCCCCC

S2399, 638-08-4, C36H70O3

Spectrum™ Stearic Anhydride
Packaging Amber Glass Bottle
Quantity 100 g
CAS Max % 1
Grade Reagent