Stearns Heavy-Duty Zipper Floatation Vest
USCG-approved Type III vest
Manufacturer: R3 Safety I416 XXL
 View more versions of this product
Catalog No. 18-999-1253E
  Check Availability
Add to Cart
    Specifications
    Certifications/Compliance USCG Type III
    Closure Type Heavy-duty zipper
    Color Orange
    Includes Reflective panels
    Size 2XL (50 to 52 in.)
    Material Nylon
    Product Type Stearns Floatation Vest
  • Description & Specifications
Related Products
Product Content Feedback