Learn More
Stearoyl-L-carnitine, ≥97.0% (HPLC), MilliporeSigma™ Supelco™
Manufacturer:  MilliporeSigma Supelco 0808450MG
 View more versions of this product
Catalog No. 11-101-3751
Add to cart
Description
Description
Specifications
Specifications
25597-09-5
(2 R)-3-Carboxy-N,N,N-trimethyl-2-[(1-oxooctadecyl)oxy]-1-propanaminium inner salt; L-Carnitine octadecanoyl ester; C18-Carnitine; Octadecanoyl-L-carnitine
≥97.0% (HPLC)
50 mg
C25H49 NO4
427.66
Analytical Standard
Analytical Standard
Safety and Handling
Safety and Handling
ShelfLife : Limited shelf life, expiry date on the label