Brady™ Steel Containment Pallet

Comply with governmental secondary containment regulations or get the security of knowing your facility is protected. Brady™ spill pallets offer an extra level of protection.

Manufacturer: Brady™ SS4DR

 View more versions of this product

Catalog No. 19-103-124

  Check Availability
Add to Cart

    Specifications

    Color Silver
    For Use With (Equipment) Four drums
    Quantity 1
    Material Galvanized steel

    • Whether you are looking to comply with governmental secondary containment regulations or just want the extra security of knowing your facility is protected, spill pallets offer an extra level of protection
    • Offers ample capacity for containing potentially hazardous leaks
    • Galvanized steel build is corrosion resistant and forklift compatible

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit