Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Steel Grip Over End Grain Leather Finger Guards

Over end finger guards constructed of grain leather offer individual finger protection.

$138.00

Specifications

Color Natural
Description Over End
For Use With (Application) Assembly, Buffing, Grinding, Sanding Operations, and Wire Pulling
Material Grain Leather
Product Type Specs Finger Guard
View More Specs

Products 2
Catalog Number Mfr. No. Size Price Quantity & Availability  
Catalog Number Mfr. No. Size Price Quantity & Availability  
19-819-581
View Documents
Steel Grip
15958 L
Large
Pack of 100 for $138.00
Add to cart
 
19-087-099
View Documents
Steel Grip
15922 M
Medium This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000. N/A
Description

Description

  • Over End Finger Guard
  • Grain Leather
Specifications

Specifications

Natural
Assembly, Buffing, Grinding, Sanding Operations, and Wire Pulling
Finger Guard
Over End
Grain Leather
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit