Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Steel Grip Flame-Resistant Cotton Coveralls

Manufacturer:  Steel Grip GS 16700MEDIUM

 View more versions of this product

Catalog No. 19-080756

This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000.

Alert:

Caution! Not for use as fire protection; avoid exposing coveralls to open flames or intense heat.


Description

Description

Comfortable, lightweight 9 oz. cotton with sateen finish. Proban™ FR-7A™ treatment imparts flame resistance. Primary protection for low heat and minor spark operations; secondary protection in high heat or open flame situations. Feature inside left breast pocket and two back and two front exterior pockets. Side slits for easy access to clothing beneath. Snaps in front and at each wrist. Standard two-piece laydown collar. Proportional inseams. Green.

Specifications

Specifications

Safety
Lay down
34 in.
Sateen FR Cotton
5
40 to 42 in.
71.12cm
Snap
Green
86.36cm
Collared
Medium
28 in.
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit