Learn More
Steel Grip Flame-Resistant Welding Jackets
Provides primary protection in low heat and minor spark operations; secondary protection in high heat, open flame, or similar situations
Manufacturer:  Steel Grip GS 167502XLARGE
 View more versions of this product
Catalog No. 19-052119
Add to cart
Alert:
Not for use as fire protection; avoid exposing jacket to open flames or intense heat.
Description
Description
Same fabric as flame-resistant cotton coveralls in a 30 in. long (76cm) jacket. Proban™ FR-7A treatment imparts flame resistance. Provides primary protection in low heat and minor spark operations; secondary protection in high heat, open flame, or similar situations. Features snap closures on front and both wrists. Set-in sleeves, 3 in. (8cm) laydown collar. Inside left breast pocket only. Proportional sleeve length. Green.
Specifications
Specifications
Safety
3 in. Lay down
Green
Sateen FR Cotton
30 in.
76.2cm
52 to 54 in.
Snap
NFPA 701
2X-Large
36 in.
91.44cm
1.0