Learn More
Steel Grip Nylon Arm Guards
Guards provide protection from cuts and lacerations caused by glass, sheet or scrap metal and other sharp objects
Manufacturer:  Steel Grip CN 8209SMALL
 View more versions of this product
Catalog No. 19-819-651
Add to cart
Alert:
Not for high-heat applications.
Description
Description
  • Lightweight 2-ply nylon
  • Adjustable Velcro™ closure
To figure your size, measure the circumference of the wrist and forearm.
Specifications
Specifications
Hook and loop
9 in.
Nylon
16.5cm
White
22.8cm
Small