Learn More
Steel Grip Spats
Offer heat and spark protection to the upper foot and ankle
Manufacturer:  Steel Grip CL 374 H
Catalog No. 19-055117
Add to cart
Description
Description
7 in. high (17.8cm), split leather spats are primarily designed to protect wearer from radiant heat, sparks, and molten metal. Pearl gray.
  • 2 in. (5.1cm) elastic band at top with snap closure
  • 6 in. (15.2cm) flexo split leather flare
  • Two rear overshoe buckle fasteners
  • Permanently attached instep strap
Specifications
Specifications
Safety
7 in.
Leather
Gray
Elastic band at top with snap closure, flexo split Leather flare, 2 rear overshoe buckle fasteners, attached instep strap