Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Steel Grip Open End Split Leather Finger Guards
SDP

Click to view available options
Size:
Medium
Large
This item is not returnable. View return policy
This item is not returnable. View return policy

Open end finger guards constructed of split leather offer individual finger protection.

  • Open End Finger Guard
  • Split Leather
Description Open End
Color Natural
Disposable Yes
Material Split Leather
Product Type Finger Guard
Size Medium
For Use With (Application) Assembly, Buffing, Grinding, Sanding Operations, and Wire Pulling
Length (English) 3.5 in.
Length (Metric) 8.9 cm