Steel Grip™ Steamgard™ 550 Protective-Steam Coveralls
Manufacturer: Steel Grip Inc. S550CL2
 View more versions of this product
Catalog No. 19-819-724
  Check Availability
Add to Cart
    Specifications
    Product Type Coveralls, Liner
    Size Medium
    For Use With (Application) Provides extended protection against steam exposure to 550 psig and 477°F
    Construction Alumized Kevlar™
  • Description & Specifications
Related Products
Product Content Feedback