Learn More
Steel Grip™ Steamgard™ 550 Protective-Steam Coveralls
Manufacturer:  Steel Grip Inc. S550CL3
 View more versions of this product
Catalog No. 19-819-725
  Check Availability
Add to cart
Specifications Description & Specifications
Specifications
Alumized Kevlar™
Large
Provides extended protection against steam exposure to 550 psig and 477°F
Coveralls, Liner