Learn More
Steel Grip Steamgard™ 550 Protective-Steam Gloves
Manufacturer: Steel Grip S550GX3
This item is not returnable.
View return policy
Specifications
Provides extended protection against steam exposure to 550 psig and 477°F | |
X-Large | |
Protective-Steam Gloves |