Steel Grip™ Steamgard™ 550 Protective-Steam Hood Liner
Manufacturer: Steel Grip Inc. S550HL2
 View more versions of this product
Catalog No. 19-819-732
  Check Availability
Add to Cart
    Specifications
    For Use With (Application) Provides extended protection against steam exposure to 550 psig and 477°F
    Size Medium
    Construction Alumized Kevlar™
    Weight (English) 7 oz.
  • Description & Specifications
Related Products
Product Content Feedback