Learn More
Steel Grip Thermonal Gloves
Special heat resistant tan split leather palm, finger & thumb reinforcement
Manufacturer:  Steel Grip TH 22514 F
Catalog No. 19-819-738
Add to cart
Description
Description
  • 22 oz. Thermonol
  • 11 oz. Wool lining
  • 14 in. Length, one size only
  • Clute cut, set in thumb style
  • Kevlar thread sewn
Specifications
Specifications
Clute cut
Wool
623.68g
14 in.
Wool
Universal
22 oz.
35.56cm