Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

edsal Steel Shelving Products for Science Education

For offices, stores, shops, basement garages and storerooms. Steel Shelving is used where easy storage is required.

Manufacturer:  edsal 29185CL

 View more versions of this product

Catalog No. S51142CMF


Add to cart

Description

Description

  • 22 gauge shelf holds up to 181.4kg (400 lb.)
  • Permanent baked enamel finish is fully rust resistant
  • Posts are punched at 2.54cm (1 in.) intervals for convenient shelf adjustments
  • Each unit is complete and can be moved independently
  • Shelves can be adjusted without disturbing shelves in adjoining units
  • Edsal's simple-to-assemble parts convert shelving to fit any storage needs
  • KwikKlip assembly for fast easy assembly or adjustment
Specifications

Specifications

Gray
45.72cm
75 in.
400 lb.
5
Close
91.44cm
18 in.
For offices, stores, shops, basement garages, storerooms
190.5cm
181.43kg
22G Steel
36 in.
SDS
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit