Alfa Aesar™ Steel shot, non-spherical, 0.25mm (0.0098 in.) dia.

Manufacturer: Alfa Aesar™ 44234A3

Quantity 2kg

 View more versions of this product

Catalog No. AA44234A3

  Check Availability
Add to Cart

    Chemical Identifiers

    MDL Number MFCD00678174
    Chemical Name or Material Steel shot, 0.25mm (0.0098 in.) dia.

    Specifications

    Physical Form Non-Spherical
    Quantity 2kg
    TSCA TSCA
    MDL Number MFCD00678174
    Chemical Name or Material Steel shot, 0.25mm (0.0098 in.) dia.

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit