Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Stella/Dppa3 Antibody - BSA Free, Novus Biologicals™

Rabbit Polyclonal Antibody
$196.10 - $499.50
Specifications
Antigen | Stella/Dppa3 |
---|---|
Concentration | 1 mg/mL |
Applications | Western Blot, ELISA |
Classification | Polyclonal |
Conjugate | Unconjugated |
Description
Stella/Dppa3 Polyclonal specifically detects Stella/Dppa3 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA.Specifications
Stella/Dppa3 | |
Western Blot, ELISA | |
Unconjugated | |
RUO | |
Human, Mouse, Rat | |
developmental pluripotency associated 3, DPPA3, DPPA3 developmental pluripotency associated 3 | |
DPPA3 | |
IgG | |
Affinity Purified |
1 mg/mL | |
Polyclonal | |
Rabbit | |
Developmental Biology | |
PBS with 0.02% Sodium Azide | |
359787 | |
Antibody was raised against a 13 amino acid synthetic peptide from near the carboxy terminus of human Stella. The immunogen is located within the last 50 amino acids of Stella. Amino Acid Squence: NGWDPSENARIGN | |
Primary |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title