Learn More
PicoStudio STEM+ Wind Competition Products for Science Education
The STEM+ Wind Competition provides students with the opportunity to see who can build the most efficient wind turbine.
$199.99
Specifications
Product Type STEM Kit
View More Specs
Alert:
Small Parts - Not recommended for children under 3 years old.
Products 2
Catalog Number Mfr. No. Description Includes Price Quantity & Availability  
Catalog Number Mfr. No. Description Includes Price Quantity & Availability  
S4131913 Products for Science Education
View Documents
PicoStudio
PS30061
Hybrid 2 Vertical Turbines, 2 Horizontal Turbines
Each for $199.99
Add to cart
S4131914 Products for Science Education
View Documents
PicoStudio
PS30062
Vertical Turbines 4 Vertical Turbines
Each for $199.99
Add to cart
Description
Description
  • Take STEM learning to the next level by adding the spirit of competition to the equation.
  • Each STEM Plus Interface allows up to 4 alternating power sources to be compared at one time.
  • Using a computer, students can see real-time data comparison or RPMs and power output of their alternators.
  • Students can compete with each other to see who can build the most efficient wind turbine.
  • With the option to construct both Horizontal and Vertical Wind Turbines, students can experience two different types of AC wind turbines.
  • This kit contains all parts needed to construct 4 wind turbines, and a STEM Plus Playoff Interface board.
Specifications
Specifications
STEM Kit