Stemi™ DV4 Series Stereomicroscopes with LED Illumination
Economical models with excellent resolution
Manufacturer: Carl Zeiss, Inc. 4354219901000
 View more versions of this product
Catalog No. 12-070-850
 N/A
N/A
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. 
    Specifications
    Base Style C stand
    Magnification Power 8x to 32x
    Objective Types 8x to 32.0x
    Quantity 1
    Eyepiece 10X 20mm widefield
    Lighting Type LED
    Zoom Range 4:1
    Includes Stand C, LED illumination, mains input plug EU, GB, USA/J, AUS
  • Description & Specifications
Related Products
Product Content Feedback