Learn More
Gibco™ StemPro™ Neural Stem Cells Non-distribution product as customer accommodation.
StemPro™ Neural Stem Cells
Manufacturer:  Gibco™ A15654
 View more versions of this product
Catalog No. A15654
Add to cart
Description
Description
StemPro™ Neural Stem Cells are cryopreserved human fetal brain-derived neural stem cells (NSCs). These cells are manufactured under Good Manufacturing Practice (GMP) manufacturing standards in a California-licensed facility and are available for Research Use Only. They are isolated and expanded in culture to passage 4 under a low oxygen (ischemic tolerant,) proprietary manufacturing process using FBS-supplemented media and then cryopreserved in an animal origin-free cryopreservation medium. A Certificate of Analysis is available for each lot that documents the quality control specifications, test results, and donor information. These cells have no observed tumorigenicity/toxicity in GLP-compliant animal studies.
StemPro™ Neural Stem Cells help you to:
  • Minimize assay variability with itNSCs manufactured using GMP manufacturing standards
  • Improve functionality studies with immature, high potency NSCs
  • Complete experiments requiring one large lot of NSCs
  • Minimize your chance of assay failure by using tested cells, reagents, and protocols
    Immature, High Potency, Low Oxygen-Cultured NSCs
    StemPro™ Neural Stem Cells are derived from human fetal brain from qualified, traceable donors. The cells are isolated, cultured, and expanded under Good Manufacturing Practice (GMP) manufacturing standards in a California-licensed facility using a proprietary Reduced Oxygen Tension manufacturing process. Manufacture of cells in a reduced oxygen tension environment results in higher yields of highly potent immature stem cells compared to cells expanded in normal oxygen culture conditions.
    Safety Tested
    StemPro™ Neural Stem Cells have been demonstrated to be non-toxic and non-tumorigenic in GLP animal studies performed according to FDA guidelines.
    Large lots can be made available for those requiring assay standardization. Protocols using tested reagents are provided to help minimize your chance of assay failure using reagents and cells that have not been evaluated to work together. View our tested media and reagents.
    High Post-Cryopreservation Viability
    These StemPro™ Neural Stem Cells are supplied in CryoStor™ cryopreservation medium at a concentration of 5x106 viable cells/vial. CryoStor™ medium is animal origin-free and shown to support high cell viabilities post-thawing. Controlled processes are used for cryopreservation, handling, and shipping to help minimize risk of adverse effects to the quality of the cells.
    Made in USA by Stemedica for Thermo Fisher Scientific.
  • Specifications
    Specifications
    StemPro™ Neural Stem Cells, 1 Vial, 1 x 106 Cells Stem Cells (Neural) Cell Type, Fetal, FBS Supplemented, Normal Donor Attributes, Cryopreserved, StemPro™ NSC SFM Media Recommendation, Human Species, GMP Certified, Spheroid Culture Type
    Liquid Nitrogen
    1 x 106 cells
    Stem cell culture
    Neural Cells
    Documents
    Provide Content Correction
    The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
    Product Title
    Cancel Submit
    For Research Use Only. Not for use in diagnostic procedures.