Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Gibco™ StemPro™ Neural Stem Cells Non-distribution product as customer accommodation.

StemPro™ Neural Stem Cells

$600.00

Specifications

Quantity 1 x 106 cells
Storage Liquid Nitrogen
Cell Type Neural Cells
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
A15654 Non-distribution product as customer accommodation. View Documents Gibco™
A15654
1 x 106 cells Each for $600.00
Add to cart
 
Description

Description

StemPro™ Neural Stem Cells are cryopreserved human fetal brain-derived neural stem cells (NSCs). These cells are manufactured under Good Manufacturing Practice (GMP) manufacturing standards in a California-licensed facility and are available for Research Use Only. They are isolated and expanded in culture to passage 4 under a low oxygen (ischemic tolerant,) proprietary manufacturing process using FBS-supplemented media and then cryopreserved in an animal origin-free cryopreservation medium. A Certificate of Analysis is available for each lot that documents the quality control specifications, test results, and donor information. These cells have no observed tumorigenicity/toxicity in GLP-compliant animal studies.

StemPro™ Neural Stem Cells help you to:

  • Minimize assay variability with itNSCs manufactured using GMP manufacturing standards
  • Improve functionality studies with immature, high potency NSCs
  • Complete experiments requiring one large lot of NSCs
  • Minimize your chance of assay failure by using tested cells, reagents, and protocols

    Immature, High Potency, Low Oxygen-Cultured NSCs
    StemPro™ Neural Stem Cells are derived from human fetal brain from qualified, traceable donors. The cells are isolated, cultured, and expanded under Good Manufacturing Practice (GMP) manufacturing standards in a California-licensed facility using a proprietary Reduced Oxygen Tension manufacturing process. Manufacture of cells in a reduced oxygen tension environment results in higher yields of highly potent immature stem cells compared to cells expanded in normal oxygen culture conditions.

    Safety Tested
    StemPro™ Neural Stem Cells have been demonstrated to be non-toxic and non-tumorigenic in GLP animal studies performed according to FDA guidelines.

    Large lots can be made available for those requiring assay standardization. Protocols using tested reagents are provided to help minimize your chance of assay failure using reagents and cells that have not been evaluated to work together. View our tested media and reagents.

    High Post-Cryopreservation Viability
    These StemPro™ Neural Stem Cells are supplied in CryoStor™ cryopreservation medium at a concentration of 5x106 viable cells/vial. CryoStor™ medium is animal origin-free and shown to support high cell viabilities post-thawing. Controlled processes are used for cryopreservation, handling, and shipping to help minimize risk of adverse effects to the quality of the cells.

    Made in USA by Stemedica for Thermo Fisher Scientific.
  • Specifications

    Specifications

    1 x 106 cells
    Liquid Nitrogen
    Neural Cells

    For Research Use Only. Not for use in diagnostic procedures.

    Provide Content Correction

    The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

    Product Title

    By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

    Cancel Submit