Learn More
GAMMASUPPLIES STER NIT/PCP SYS 10IN PORT 20C
Supplier:  GAMMASUPPLIES GSG10NIT85
Encompass_Preferred
Sterile Nitrile/PCP Sleeve/Glove System for Isolators/Glove Boxes; Meet USP 797; Rated for 8 hours against cytotoxins; 10 in. Port (Circumference 32 in./800mm); ISO 11137 SAL 10-6 ; IEST-RP-CC 005.3; by gammaSUPPLIES; packaged individually; batch/lot #, expire
Catalog No. 50-144-004
Please call customer service for current pricing. \n1-800-766-7000