Learn More
AdipoGen Sterigmatocystin, 10048-13-2, MFCD32182631, 5 mg
SDP
Supplier:  AdipoGen BVT0171M005
Encompass_Preferred
MF: C18H12O6, MW: 324.3, ≥98%, Appearance: Pale yellow powder. Intermediate of the biosynthetic pathway to aflatoxin B1. Mycotoxin. DNA synthesis inhibitor. Anticancer compound. Cytotoxic, carcinogenic and mutagenic. Acyl-CoA:cholesterol acyltransferase 2 (ACAT2) inhibitor. Shown to induce apoptosis in human peripheral lymphocytes and necrosis in rat liver. Induces sister chromatid exchanges in murine bone marrow cells.
Catalog No. 50-595-565
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.