Thermo Scientific™ Automation Tips for Tecan™ Liquid Handling Platforms with Liquid Handling (LiHa) Arm Available on GSA/VA Contract for Federal Government customers only.

Increase pipetting performance with Thermo Scientific™ Automation Tips for Tecan™ Liquid Handling Platforms, which undergo a rigorous 15-point quality control program to ensure consistency.

Manufacturer: Thermo Scientific™ 901261

 View more versions of this product

Catalog No. 21-402-331

  Check Availability
Add to Cart

Disclaimers:

Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.


    Specifications

    Color Clear
    For Use With (Equipment) Carvro
    Volume (Metric) 20μL,20µL
    Sterility Sterile
    Packaging 96 Tips/Tray, 10 Trays/Case,96 Tips/Tray, 10 Trays/Case
    Filtered No
    Sterile Yes
    Features Standard
    Item Description 901-261
    Automated Workstation Brand Tecan
    Automated Workstation Model Genesis, Freedom Evo, MiniPrep and Carvro

    Increase pipetting performance with Thermo Scientific™ Automation Tips for Tecan™ Liquid Handling Platforms, which undergo a rigorous 15-point quality control program to ensure consistency. Their superior tip straightness and a low coefficient of variation provide accurate and precise performance. Compatible with: Tecan Cavro, Genesis, MiniPrep and Freedom EVO; PerkinElmer/Packard MultiPROBE II HT, PerkinElmer/Packard MultiPROBE II HT EX and PerkinElmer JANUS.

    • Available as standard, filtered and/or liquid sensing
    • Certified free of RNase, DNase, DNA, endotoxin, bioburden and pyrogen
    • Lot-tested during production on actual workstation for which they are created
    • Fully conductive, providing increased pipetting performance
    • Low percentage coefficient of variation (%CV)

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit