Learn More
High-Tech Conversions Sterile Dry Cleanroom Wipes
High-Tech Conversions Sterile Dry Cleanroom Wipes meet USP 797 and 800 requirements for critical contamination control
Specifications
Autoclavable No
Basis Weight 68 gsm
Certifications/Compliance ANSI/AAMI/ISO 11137:2013, ISO Class 5-6 (100-1000)
Color White
Disposable Yes
View More Specs
Products 2
Catalog Number Mfr. No. Dimensions (L x W) Price Quantity & Availability  
Catalog Number Mfr. No. Dimensions (L x W) Price Quantity & Availability  
50-100-2363
View Documents
High-Tech Conversions
NT1099CB/DEL
9 x 9 in. This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000. View substitute products »
50-100-2364
View Documents
High-Tech Conversions
NT101212CB/DEL
12 x 12 in. This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000. View substitute products »
Description
Description
  • Validated Sterile
  • ISO Cleanroom 5-6
  • Poly-Cellulose Blend
  • Cobalt-60 Gamma Irradiated
  • Meets USP 797 and 800 requirements for critical contamination control
Specifications
Specifications
No
ANSI/AAMI/ISO 11137:2013, ISO Class 5-6 (100-1000)
Yes
Poly-Cellulose Blend
Sterile
68 gsm
White
Wiping/Cleaning all aspects of Class 5-8 Cleanrooms
Non-Woven, Sterile Cleanroom Wipes