Learn More
Kimberly-Clark Professional™ Sterile Evolution™ 4 Surgical Gowns
Fourth-generation fabric technology provides advanced protection, breathability and comfort
Manufacturer:  Kimberly-Clark Professional™ 90012
Catalog No. 19-077-549
Add to cart
Description
Description
  • Raglan sleeves
  • Adjustable neckline with yellow neck binding - hook and loop
  • Drop fold
  • Expected Fluid Contact: Low to Moderate
  • Flame Resistance: Meets NFPA Class I
  • Comes with Towel
  • Sterile
  • Comes one per pack; 36 per case
Specifications
Specifications
Safety